EMAIL THIS PAGE TO A FRIEND
81114 Sigma-Aldrich

GC Stationary Phase

phase Synperonic PE/L64, bottle of 10 mL

Purchase

Properties

Related Categories Analytical/Chromatography, Biochemicals and Reagents, Detergents, Detergents N to Z, GC Stationary Phases,
grade   liquid phase for GC
description   block copolymer of polyethylene and polypropylene glycol
  non-ionic
mol wt   mol wt <2900
packaging   bottle of 10 mL
application(s)   gas chromatography (GC): suitable
matrix active group   Synperonic PE/L64 phase
density   1.05 g/mL at 20 °C

Description

General description

Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MD0911050

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?