• USA Home
  • 81129 - Poly(acrylic acid sodium salt)

EMAIL THIS PAGE TO A FRIEND
81129 Sigma-Aldrich

Poly(acrylic acid sodium salt)

for GPC, standard, 62,900

Synonym: Poly(sodium acrylate), Sodium polyacrylate

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Poly (acrylic acid sodium salt), Polymer Standards,
grade   for GPC
  standard
form   neat
CofA   exact values can be taken from the accompanying certificate of analysis
mol wt   Mn ~47900
  Mp ~62900
  Mw ~83400
application(s)   gel permeation chromatography (GPC): suitable
Mw/Mn   ~1.74
format   neat

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
WD6826000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?