• USA Home
  • 81297 - Poly(ethylene glycol)

EMAIL THIS PAGE TO A FRIEND
81297 Sigma-Aldrich

Poly(ethylene glycol)

standard, for GPC, 17,500

Synonym: PEG

Purchase

Properties

grade   for GPC
  standard
vapor density   >1 (vs air)
vapor pressure   <0.01 mmHg ( 20 °C)
form   neat
CofA   exact values can be taken from the accompanying certificate of analysis
autoignition temp.   581 °F
mol wt   Mn ~12800
  Mp ~16000
  Mw ~14100
application(s)   gel permeation chromatography (GPC): suitable
Mw/Mn   ~1.10
format   neat

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TQ3500000
Flash Point(F) 
444.2 °F
Flash Point(C) 
229 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?