• USA Home
  • 81316 - Poly(ethylene glycol) methyl ether

EMAIL THIS PAGE TO A FRIEND
81316 Sigma-Aldrich

Poly(ethylene glycol) methyl ether

average Mn 500

Synonym: Methoxy poly(ethylene glycol), Polyethylene glycol monomethyl ether, mPEG

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents N to Z, Non-Ionic Detergents
vapor density   >1 (vs air)
vapor pressure   0.05 mmHg ( 20 °C)
description   non-ionic
mol wt   average Mn 500
  mol wt 470-530
mp   20 °C(lit.)

Description

Other Notes

Catalyst for phase-transfer reactions; Preparation of monofunctional PEGs

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
359.6 °F
Flash Point(C) 
182 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?