• USA Home
  • 81318 - Poly(ethylene glycol) methyl ether

EMAIL THIS PAGE TO A FRIEND
81318 Sigma-Aldrich

Poly(ethylene glycol) methyl ether

350

Synonym: Methoxy poly(ethylene glycol), Polyethylene glycol monomethyl ether, mPEG

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents N to Z, Non-Ionic Detergents
vapor density   >1 (vs air)
vapor pressure   0.05 mmHg ( 20 °C)
description   non-ionic
form   liquid
  viscous liquid
mol wt   Mw 363 (D)
refractive index   n20/D 1.457
mp   −8 °C(lit.)
density   1.088 g/mL at 20 °C
functional group   OH

Description

Other Notes

This product has been replaced by M6768-SIGMA | Methoxypolyethylene glycol 350 average mol wt 350

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
PE8271000
Flash Point(F) 
359.6 °F
Flash Point(C) 
182 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M6768 Methoxypolyethylene glycol 350, average mol wt 350

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?