• USA Home
  • 81321 - Poly(ethylene glycol) methyl ether

EMAIL THIS PAGE TO A FRIEND
81321 Sigma-Aldrich

Poly(ethylene glycol) methyl ether

average Mw 2,000

Synonym: Methoxy poly(ethylene glycol), Polyethylene glycol monomethyl ether, mPEG

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents N to Z, Hydroxy, Materials Science,
vapor density   >1 (vs air)
vapor pressure   0.05 mmHg ( 20 °C)
description   non-ionic
form   flakes
  powder or crystals
mol wt   average Mw 2,000
mp   52-56 °C
Ω-end   OH
α-end   CH3

Description

Price and Availability


Laboratory Gloves

The Future of Bioimaging
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
PE8271000
Flash Point(F) 
359.6 °F
Flash Point(C) 
182 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

Degradable Poly(ethylene glycol) Hydrogels for 2D and 3D Cell Culture

Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. For decades, research in polymeric biomateria...
Keywords: Absorption, Angiogenesis, Biomaterials, Catalysis, Cell culture, Degradations, Eliminations, Growth factors, Infrared spectroscopy, Ligands, Michael Addition, Polymerization reactions, Ring-opening polymerization

Fouling Resistant Biomimetic Poly(Ethylene Glycol) Based Grafted Polymer Coatings

Accumulation of biological matter at surfaces is an inevitable event in virtually any environment in which natural and man-made materials are used. Although sometimes fouling of surfaces with biomole...
Lesley M. Hamming and Prof. Phillip B. Messersmith
Material Matters 2008, 3.3, 52.
Keywords: Adhesion, Adsorption, Biomaterials, Cell attachment, Derivatizations, Electrophoresis, Immobilization, Methods, Polymerization reactions, Radical polymerization, Spectroscopy, Surface Science, Tools, Type

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?