• USA Home
  • 81322 - Poly(dibenzo-18-crown-6)

EMAIL THIS PAGE TO A FRIEND
81322 Sigma-Aldrich

Poly(dibenzo-18-crown-6)

purum, ≥97.0% (CH)

Purchase

Properties

Related Categories Chelation/Complexation Compounds, Chemical Synthesis, Crown Ethers, Synthetic Reagents
grade   purum
assay   ≥97.0% (CH)

Description

Other Notes

Dibenzo-18-crown-6 condensation resin useful for an array of reactions; Used in analysis for the adsorption of metals from solutions

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?