• USA Home
  • 81345 - Trimethylsilyl polyphosphate

EMAIL THIS PAGE TO A FRIEND
81345 Sigma-Aldrich

Trimethylsilyl polyphosphate

Synonym: PPSE, Polyphosphoric acid trimethylsilyl ester

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Inorganic Acids, Materials Science,
InChI Key   YSVQLWJDHYBITL-UHFFFAOYSA-N
density   1.18 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Versatile reagent used for many reactions involving dehydration; e.g. substitutions, rearrangements, condensations

This product has been replaced by 414026-ALDRICH | Trimethylsilyl polyphosphate

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
3
Flash Point(F) 
39.2 °F
Flash Point(C) 
4 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

414026 Trimethylsilyl polyphosphate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?