EMAIL THIS PAGE TO A FRIEND
81374 Sigma-Aldrich

Poly(vinyl alcohol)

standard, for GPC, Mw 122,400

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Organic Soluble Polymers, Poly(vinyl alcohol),
grade   for GPC
  standard
form   neat
CofA   exact values can be taken from the accompanying certificate of analysis
mol wt   Mn ~60100
  Mp ~85700
  Mw ~122400
application(s)   gel permeation chromatography (GPC): suitable
Mw/Mn   ~2.04
format   neat

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
TR8100000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?