• USA Home
  • 81496 - Poly(methyl methacrylate)

EMAIL THIS PAGE TO A FRIEND
81496 Sigma-Aldrich

Poly(methyl methacrylate)

standard, for GPC, 5,000

Synonym: PMMA, Poly(methacrylic acid methyl ester)

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Organic Soluble Polymers, Poly(methyl methacrylate),
grade   for GPC
  standard
form   neat
CofA   exact values can be taken from the accompanying certificate of analysis
autoignition temp.   580 °F
mol wt   Mn ~4760
  Mp ~5090
  Mw ~5050
application(s)   gel permeation chromatography (GPC): suitable
solubility   H2O: insoluble
density   1.200 g/cm3
Mw/Mn   ~1.06
format   neat

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
TR0400000
Flash Point(F) 
>482 °F
Flash Point(C) 
>250 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?