EMAIL THIS PAGE TO A FRIEND
81552 Sigma-Aldrich

Prednisone

tested according to Ph.Eur.

Synonym: 1,4-Pregnadiene-17α,21-diol-3,11,20-trione, 1-Cortisone, 17α,21-Dihydroxy-1,4-pregnadiene-3,11,20-trione, Dehydrocortisone, Prednisonum

  • CAS Number 53-03-2

  • Empirical Formula (Hill Notation) C21H26O5

  • Molecular Weight 358.43

  •  Beilstein Registry Number 2065301

  •  EC Number 200-160-3

  •  MDL number MFCD00003608

  •  PubChem Substance ID 329769211

Purchase

Properties

Related Categories Analytical/Chromatography, Core Bioreagents, Enzyme Inhibitors, Pharmacopoeia, Pharmacopoeia A-Z,
InChI Key   XOFYZVNMUHMLCC-ZPOLXVRWSA-N
pharmacopeia   testing conforms to Pharmacopeia
mp   236-238 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TU4154100

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?