• USA Home
  • 81605 - Poly(styrenesulfonic acid sodium salt)

EMAIL THIS PAGE TO A FRIEND
81605 Sigma-Aldrich

Poly(styrenesulfonic acid sodium salt)

standard, for GPC, 1,400

Synonym: Poly(sodium styrenesulfonate)

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Poly(styreneslfonic acid sodium salt), Polymer Standards,
grade   for GPC
  standard
form   neat
CofA   exact values can be taken from the accompanying certificate of analysis
mol wt   Mp ~1370
  Mw ~1430
application(s)   gel permeation chromatography (GPC): suitable
Mw/Mn   ~1.2
format   neat

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DB6836000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?