EMAIL THIS PAGE TO A FRIEND
81648 Sigma-Aldrich

Pril® Mannitol Agar

for microbiology

Purchase

Properties

Related Categories All Food Control Media (alphabetical), All Solid Media/Agars, Clinical Media, Media, Media by Application,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   agar, 13.0 g/L
  disodium hydrogen phosphate, 2.0 g/L
  D-mannitol, 15 g/L
  meat extract, 7.0 g/L
  meat peptone, 10.0 g/L
  metachrome yellow, 1.875 g/L
  Pril®, 2.0 g/L
  sodium chloride, 3.0 g/L
  water blue, 0.625 g/L
final pH   7.2±0.2 (25 °C)
suitability   selective and differential for Salmonella

Description

Application

Selective agar for the isolation and differentiation of Salmonella species alongside Proteus acc. to Pietzsch.

Other Notes

In Salmonella diagnostics

Legal Information

Pril is a registered trademark of Henkel KGaA

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Differentiation and Identification Media for Salmonella

Salmonella contamination is the second leading cause of food-borne illness worldwide. Controlling outbreaks of Salmonella is an important task for food regulators, restaurants and the food industry i...
Jvo Siegrist
Microbiology Focus Edition 1.3
Keywords: Biochemistry, Fermentation, Food & Beverage, Indicators, Microbiology

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?