• USA Home
  • 81662 - Prionex® protein stabilizer solution from hog collagen

EMAIL THIS PAGE TO A FRIEND
81662 Sigma-Aldrich

Prionex® protein stabilizer solution from hog collagen

sterile, 10% (in water)

Synonym: Protein stabilizer Prionex®

Purchase

Properties

Related Categories Biochemicals and Reagents, Biostabilization Solutions, Protein Stabilizers, Stabilization of Biomolecules
assay   10% (in water)
quality   sterile
impurities   1.3-1.9% N (dried substance)
pH   6.0-7.5

Description

Other Notes

Sales restrictions may apply

Same product — same quality — now Sigma-Aldrich brand

Legal Information

Prionex is a registered trademark of Pentapharm AG, Basel

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?