• USA Home
  • 81740 - L-Proline methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
81740 Sigma-Aldrich

L-Proline methyl ester hydrochloride

puriss., ≥99.0% (AT)

  • CAS Number 2133-40-6

  • Empirical Formula (Hill Notation) C6H11NO2 · HCl

  • Molecular Weight 165.62

  •  Beilstein Registry Number 3596045

  •  EC Number 218-363-0

  •  MDL number MFCD00012708

  •  PubChem Substance ID 329769214

Purchase

Properties

InChI Key   HQEIPVHJHZTMDP-JEDNCBNOSA-N
grade   puriss.
assay   ≥99.0% (AT)
optical activity   [α]20/D −33±2°, c = 1% in H2O
mp   68-78 °C
  69-71 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

This product has been replaced by 287067-ALDRICH | L-Proline methyl ester hydrochloride 98%

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Proline Methyl Ester Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX® G-TA

From our library of Articles, Sigma-Aldrich presents GC Analysis of Proline Methyl Ester Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX® G-TA
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Proline Methyl Ester Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-TA

From our library of Articles, Sigma-Aldrich presents GC Analysis of Proline Methyl Ester Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-TA
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?