EMAIL THIS PAGE TO A FRIEND
81744 Sigma-Aldrich

D-Prolinol

purum, ≥98.0% (sum of enantiomers, GC)

Synonym: (R)-(−)-2-(Hydroxymethyl)pyrrolidine, (R)-(−)-2-Pyrrolidinemethanol

  • CAS Number 68832-13-3

  • Empirical Formula (Hill Notation) C5H11NO

  • Molecular Weight 101.15

  •  Beilstein Registry Number 1523669

  •  MDL number MFCD00064321

  •  PubChem Substance ID 329769215

Purchase

Properties

Related Categories Amino Acids & Derivatives, Amino Alcohols, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   HVVNJUAVDAZWCB-RXMQYKEDSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]20/D −31±2°, c = 1% in toluene
refractive index   n20/D 1.485
density   1.036 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NA 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
186.8 °F
Flash Point(C) 
86 °C
Protocols & Articles

Articles

Proline Derivatives and Analogs

Proline is a non-polar proteinogenic amino acid that forms a tertiary amide when incorporated into peptides. It does not have a hydrogen on the amide group and therefore cannot act as a hydrogen bond...
Chemfiles Volume 5 Article 12
Keywords: Adhesion, Biochemistry, Catalysis, Deposition, Hydroxylations, Isomerizations, Nucleic acid hybridization, Pharmaceutical, Reductions, Substitutions, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?