EMAIL THIS PAGE TO A FRIEND
81780 Sigma-Aldrich

1,3-Propanediol

puriss., ≥99.0% (GC)

Synonym: 1,3-Dihydroxypropane, Bio-PDO, PG, Trimethylene glycol

Purchase

Properties

Related Categories Building Blocks, Cell Biology, Chemical Synthesis, Glycerolipid Metabolism, Metabolic Pathways,
vapor pressure   0.8 mmHg ( 20 °C)
  9.8 mmHg ( 100 °C)
InChI Key   YPFDHNVEDLHUCE-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
refractive index   n20/D 1.440(lit.)
  n20/D 1.440
bp   214 °C/760 mmHg(lit.)
mp   −27 °C(lit.)
density   1.053 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TY2010000
Flash Point(F) 
>210.2 °F
Flash Point(C) 
>99 °C
Protocols & Articles

Articles

GC Analysis of Glycols and Diols on SPB®-1000

From our library of Articles, Sigma-Aldrich presents GC Analysis of Glycols and Diols on SPB®-1000
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P50404 1,3-Propanediol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?