• USA Home
  • 81804 - Sodium 1-propanesulfonate monohydrate

EMAIL THIS PAGE TO A FRIEND
81804 Sigma-Aldrich

Sodium 1-propanesulfonate monohydrate

puriss., ≥99.0% (T)

Synonym: 1-Propanesulfonic acid sodium salt monohydrate

  • CAS Number 304672-01-3

  • Linear Formula CH3CH2CH2SO3Na · H2O

  • Molecular Weight 164.16

  •  Beilstein Registry Number 3714817

  •  MDL number MFCD00167033

  •  PubChem Substance ID 329769217

Purchase

Properties

Related Categories Alkyl Sulfonates, Anionic Detergents, Biochemicals and Reagents, Building Blocks, Chemical Synthesis,
description   anionic
InChI Key   QBQVXXQXZXDEHE-UHFFFAOYSA-M
grade   puriss.
assay   ≥99.0% (T)
mp   250 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

358959 Sodium 1-propanesulfonate monohydrate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?