EMAIL THIS PAGE TO A FRIEND
81825 Sigma-Aldrich

Propargylamine

purum, ≥98.0% (GC)

Synonym: 2-Propynylamine, 3-Amino-1-propyne

Purchase

Properties

Related Categories Alkynes, Building Blocks, Chemical Synthesis, Organic Building Blocks, Terminal More...
InChI Key   JKANAVGODYYCQF-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.449(lit.)
  n20/D 1.449
bp   83 °C(lit.)
density   0.86 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3286 8(6.1)(3) / PGII
WGK Germany 
3
RTECS 
UK5250000
Flash Point(F) 
42.8 °F
Flash Point(C) 
6 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P50900 Propargylamine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?