EMAIL THIS PAGE TO A FRIEND
81860 Sigma-Aldrich

Propiolic acid

technical, ≥95% (T)

Synonym: Acetylenecarboxylic acid, Propynoic acid

  • CAS Number 471-25-0

  • Linear Formula HC≡CCOOH

  • Molecular Weight 70.05

  •  Beilstein Registry Number 878176

  •  EC Number 207-437-8

  •  MDL number MFCD00004360

  •  PubChem Substance ID 24887911

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   UORVCLMRJXCDCP-UHFFFAOYSA-N
grade   technical
assay   ≥95% (T)
refractive index   n20/D 1.431(lit.)
  n20/D 1.431
bp   102 °C/200 mmHg(lit.)
mp   16-18 °C(lit.)
density   1.135 g/mL at 20 °C
  1.138 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2920 3(8) / PGII
WGK Germany 
3
RTECS 
UD9300000
Flash Point(F) 
136.4 °F
Flash Point(C) 
58 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

P51400 Propiolic acid, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?