• USA Home
  • 81960 - tert-Butyl propionate

EMAIL THIS PAGE TO A FRIEND
81960 Sigma-Aldrich

tert-Butyl propionate

puriss., ≥98.5% (GC)

  • CAS Number 20487-40-5

  • Linear Formula CH3CH2COOC(CH3)3

  • Molecular Weight 130.18

  •  Beilstein Registry Number 1747857

  •  MDL number MFCD00009304

  •  PubChem Substance ID 329769225

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   JAELLLITIZHOGQ-UHFFFAOYSA-N
grade   puriss.
assay   ≥98.5% (GC)
refractive index   n20/D 1.393(lit.)
  n20/D 1.393
bp   118-118.5 °C(lit.)
density   0.865 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3272 3 / PGII
WGK Germany 
3
Flash Point(F) 
69.8 °F
Flash Point(C) 
21 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?