EMAIL THIS PAGE TO A FRIEND
81990 Sigma-Aldrich

Methyl propionate

purum, ≥99.0% (GC)

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C2 to C5, Carbonyl Compounds, Chemical Synthesis,
vapor density   3 (vs air)
vapor pressure   40 mmHg ( 11 °C)
  85.3 mmHg ( 25 °C)
InChI Key   RJUFJBKOKNCXHH-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
autoignition temp.   876 °F
expl. lim.   13 %
refractive index   n20/D 1.376(lit.)
  n20/D 1.377
bp   79 °C(lit.)
mp   −88 °C(lit.)
density   0.915 g/mL at 25 °C(lit.)

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1248 - class 3 - PG 2 - Methyl propionate
WGK Germany 
1
RTECS 
UF5970000
Flash Point(F) 
28.4 °F
Flash Point(C) 
-2 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?