EMAIL THIS PAGE TO A FRIEND
81992 Sigma-Aldrich

Sodium propionate

≥99.0% (NT)

Synonym: Propionic acid sodium salt

  • CAS Number 137-40-6

  • Linear Formula CH3CH2COONa

  • Molecular Weight 96.06

  •  Beilstein Registry Number 3566934

  •  EC Number 205-290-4

  •  MDL number MFCD00002759

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Salts, Chemical Synthesis, Organic Building Blocks More...
InChI Key   JXKPEJDQGNYQSM-UHFFFAOYSA-M
assay   ≥99.0% (NT)
mp   285-286 °C(lit.)
  287-289 °C
solubility   H2O: soluble0.1 g/mL, clear

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
UF7525000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?