EMAIL THIS PAGE TO A FRIEND
82050 Sigma-Aldrich

Propiophenone

purum, ≥99.0% (GC)

Synonym: Ethyl phenyl ketone

  • CAS Number 93-55-0

  • Linear Formula C6H5COC2H5

  • Molecular Weight 134.18

  •  Beilstein Registry Number 606215

  •  EC Number 202-257-6

  •  MDL number MFCD00009309

  •  PubChem Substance ID 329769230

Purchase

Properties

Related Categories Building Blocks, C9, Carbonyl Compounds, Chemical Synthesis, Ketones,
vapor pressure   1 mmHg ( 50 °C)
InChI Key   KRIOVPPHQSLHCZ-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.526(lit.)
  n20/D 1.527
bp   218 °C(lit.)
mp   17-19 °C(lit.)
density   1.009 g/mL at 20 °C
  1.009 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
UG7175000
Flash Point(F) 
210.2 °F
Flash Point(C) 
99 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?