EMAIL THIS PAGE TO A FRIEND
82087 Sigma-Aldrich

1-Propanol

B&J Brand (product of Burdick & Jackson)

Synonym: Propyl alcohol

  • CAS Number 71-23-8

  • Linear Formula CH3CH2CH2OH

  • Molecular Weight 60.10

  •  Beilstein Registry Number 1098242

  •  EC Number 200-746-9

  •  MDL number MFCD00002941

  •  PubChem Substance ID 329769234

Purchase

Properties

grade   B&J Brand (product of Burdick & Jackson)
vapor density   2.1 (vs air)
vapor pressure   10 mmHg ( 147 °C)
  14.9 mmHg ( 20 °C)
InChI Key   BDERNNFJNOPAEC-UHFFFAOYSA-N
autoignition temp.   700 °F
expl. lim.   13.7 %
refractive index   n20/D 1.384(lit.)
bp   97 °C(lit.)
mp   −127 °C(lit.)
density   0.804 g/mL at 25 °C(lit.)

Description

Other Notes

Sales restrictions may apply

Application

1-Propanol may be employed as a solvent for the synthesis of mesoporous alumina and esters.

General description

1-Propanol is a linear alcohol. It exhibits Debye-type relaxation process which resembles α-relaxation and Johari-Goldstein type β-relaxation features of supercooled liquids (nonhydrogen-bonding type). It has been tested as a substitute for fuel for various fuel cells. Its biosynthesis from glucose using Escherichia coli strain has been proposed.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1274 - class 3 - PG 2 - n-Propanol
WGK Germany 
1
RTECS 
UH8225000
Flash Point(F) 
71.6 °F
Flash Point(C) 
22 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?