EMAIL THIS PAGE TO A FRIEND
82240 Sigma-Aldrich

Chloropropanol

purum, mixture of isomers, ≥97.0% (GC)

Synonym: Propylene chlorohydrin

  • CAS Number 127-00-4

  • Linear Formula CH3CH(OH)CH2Cl+CH3CHClCH2OH

  • Molecular Weight 94.54

  •  Beilstein Registry Number 773653

  •  EC Number 204-819-6

  •  MDL number MFCD00004530

  •  PubChem Substance ID 329769240

Purchase

Properties

Related Categories Alcohols, Building Blocks, C2 to C6, Chemical Synthesis, Organic Building Blocks,
InChI Key   YYTSGNJTASLUOY-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.439(lit.)
  n20/D 1.439
bp   126-127 °C(lit.)
density   1.111 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2611 3(6.1) / PGII
WGK Germany 
3
RTECS 
UA8942000
Flash Point(F) 
125.6 °F
Flash Point(C) 
52 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

292087 1-Chloro-2-propanol, 70%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?