• USA Home
  • 82460 - 6-Propyl-2-thiouracil

EMAIL THIS PAGE TO A FRIEND
82460 Sigma-Aldrich

6-Propyl-2-thiouracil

purum, ≥98.0% (T)

Synonym: 2,3-Dihydro-6-propyl-2-thioxo-4(1H)-pyrimidinone, 4-Hydroxy-2-mercapto-6-propylpyrimidine

  • CAS Number 51-52-5

  • Empirical Formula (Hill Notation) C7H10N2OS

  • Molecular Weight 170.23

  •  Beilstein Registry Number 130039

  •  EC Number 200-103-2

  •  MDL number MFCD00006041

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Pyrimidines More...
description   enzyme inhibitor
InChI Key   KNAHARQHSZJURB-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
mp   218-220 °C(lit.)
  218-220 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YR1400000
Flash Point(F) 
572 °F
Flash Point(C) 
300 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

P3755 6-Propyl-2-thiouracil, enzyme inhibitor

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?