EMAIL THIS PAGE TO A FRIEND
82474 Sigma-Aldrich

Prostaglandin E1

≥99.0% (TLC)

Synonym: (11α,13E,15S)-11,15-Dihydroxy-9-oxoprost-13-enoic acid, Alprostadil, PGE1

  • CAS Number 745-65-3

  • Empirical Formula (Hill Notation) C20H34O5

  • Molecular Weight 354.48

  •  Beilstein Registry Number 5294062

  •  EC Number 212-017-2

  •  MDL number MFCD00077860

  •  PubChem Substance ID 329769246

Purchase

Properties

Related Categories Allium sativum (Garlic), Biochemicals and Reagents, Cell Biology, Eicosanoids (Prostaglandins, Leukotrienes), Fatty Acyls,
InChI Key   GMVPRGQOIOIIMI-DWKJAMRDSA-N
assay   ≥99.0% (TLC)
solubility   acetone: soluble50 mg/mL, clear, colorless to faintly yellow
storage temp.   −20°C
Gene Information   human ... PTGIR(5739)
mouse ... Ptger1(19216), Ptger2(19217), Ptger3(19218), Ptger4(19219)

Description

Other Notes

Enhancement of cAMP responses by protein kinase C activation.

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
GY4569800

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?