• USA Home
  • 82491 - Protein A (extracellular), immobilized on Agarose CL-4B from Staphylococcus aureus

EMAIL THIS PAGE TO A FRIEND
82491 Sigma-Aldrich

Protein A (extracellular), immobilized on Agarose CL-4B from Staphylococcus aureus

for affinity chromatography

Synonym: Protein A (extracellular)-Agarose CL-4B

Purchase

Properties

Related Categories Affinity, Agarose, Analytical/Chromatography, LPLC Separation Media, LPLC/TLC/Paper Chromatography More...
grade   for affinity chromatography
contains   0.02% thimerosal as preservative
capacity   ~30 mg/mL, gel binding capacity (human IgG)
density   1.020 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

General description

extracellular protein A is immobilized on cross-linked CNBr-activated agarose 4B

Physical form

50% suspension in 0.5 M NaCl

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?