• USA Home
  • 82495 - Proteinase K from Tritirachium album

EMAIL THIS PAGE TO A FRIEND
82495 Sigma-Aldrich

Proteinase K from Tritirachium album

powder, beige, ~290 U/mg

Synonym: Endopeptidase K

Purchase

Properties

Related Categories 3.4.x.x Peptidases, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Enzyme Class Index,
form   powder
mol wt   Mr 27000-29000
  mol wt 28.93 kDa
color   beige
solubility   H2O: soluble10 mg/mL, hazy, brown-yellow
storage temp.   2-8°C

Description

Biochem/physiol Actions

Proteinase K is a stable and highly reactive serine protease. Evidence from crystal and molecular structure studies indicates the enzyme belongs to the subtilisin family with an active-site catalytic triad (Asp39-His69-Ser224). It is stable in a broad range of environments: pH, buffer salts, detergents (SDS), and temperature. In the presence of 0.1-0.5% SDS, proteinase K retains activity and will digest a variety of proteins and nucleases in DNA preparations without compromising the integrity of the isolated DNA.

Other Notes

Solubilizes membrane-bound enzymes e.g. acetyl cholinesterase from Torpedo marmorata; Modifies creatine kinase isoenzymes

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol N-acetyl-L-tyrosine-ethylester (ATEE) per minute at pH 9.0 and 30 °C

Application

Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
Removes endotoxins that bind to cationic proteins such as lysozyme and ribonuclease A.
Reported useful for the isolation of hepatic, yeast, and mung bean mitochondria
Determination of enzyme localization on membranes
Treatment of paraffin embedded tissue sections to expose antigen binding sites for antibody labeling.
Digestion of proteins from brain tissue samples for prions in Transmissible Spongiform Encephalopathies (TSE) research.

Price and Availability


Laboratory Gloves

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P6556 Proteinase K from Tritirachium album, lyophilized powder, ≥30 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?