• USA Home
  • 82558 - Buffer standard solution according to DIN 19266

EMAIL THIS PAGE TO A FRIEND
82558 Sigma-Aldrich

Buffer standard solution according to DIN 19266

0.009 M potassium dihydrogen phosphate, 0.03 M disodium hydrogen phosphate, pH 7.413±0.010 (25 °C)

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, DIN 19266, European Community: ISO, DIN and DL 152,
shelf life   limited shelf life, expiry date on the label
concentration   0.009 M potassium dihydrogen phosphate
  0.03 M disodium hydrogen phosphate
pH   7.413±0.010 (25 °C)
density   1.002 g/mL at 20 °C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?