• USA Home
  • 82566 - Buffer solution pH 4.0 (20°C)

EMAIL THIS PAGE TO A FRIEND
82566 Sigma-Aldrich

Buffer solution pH 4.0 (20°C)

sodium azide ~0.05 %, sodium hydroxide ~0.068 M, citric acid ~0.056 M, sodium chloride ~0.044 M

Purchase

Properties

shelf life   limited shelf life, expiry date on the label
contains   NaN3 as stabilizer
composition   citric acid, ~0.056 M
  sodium azide, ~0.05%
  sodium chloride, ~0.044 M
  sodium hydroxide, ~0.068 M
density   1.007 g/mL at 20 °C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?