• USA Home
  • 82567 - Buffer solution pH 5.0 (20 °C)

EMAIL THIS PAGE TO A FRIEND
82567 Sigma-Aldrich

Buffer solution pH 5.0 (20 °C)

citric acid ~0.096 M, sodium hydroxide ~0.20 M

Synonym: Buffer solution from sodium citrate, Sodium citrate buffer solution

Purchase

Properties

InChI Key   HRXKRNGNAMMEHJ-UHFFFAOYSA-K
shelf life   limited shelf life, expiry date on the label
composition   citric acid, ~0.096 M
  sodium hydroxide, ~0.20 M
density   1.000 g/mL at 20 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1824 8 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?