EMAIL THIS PAGE TO A FRIEND
82620 Sigma-Aldrich

Pyrazole

purum, ≥98.0% (GC)

Synonym: 1,2-Diazole

  • CAS Number 288-13-1

  • Empirical Formula (Hill Notation) C3H4N2

  • Molecular Weight 68.08

  •  Beilstein Registry Number 103775

  •  EC Number 206-017-1

  •  MDL number MFCD00005234

  •  PubChem Substance ID 329769255

Purchase

Properties

Related Categories Building Blocks, C3 to C5, Chemical Synthesis, Heterocyclic Building Blocks, Pyrazoles More...
InChI Key   WTKZEGDFNFYCGP-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   186-188 °C(lit.)
mp   66-70 °C
  67-70 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
UQ4900000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

P56607 Pyrazole, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?