EMAIL THIS PAGE TO A FRIEND
82650 Sigma-Aldrich

Pyrene

purum, for fluorescence, ≥97.0% (GC)

Synonym: Benzo[def]phenanthrene

  • CAS Number 129-00-0

  • Empirical Formula (Hill Notation) C16H10

  • Molecular Weight 202.25

  •  Beilstein Registry Number 1307225

  •  EC Number 204-927-3

  •  MDL number MFCD00004136

  •  PubChem Substance ID 329769257

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Fluorescent Probes, Labels, Particles and Stains, Fluorescent Stains, Luminescent Compounds/Detection,
grade   for fluorescence
InChI Key   BBEAQIROQSPTKN-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
mp   145-148 °C(lit.)
  149-151 °C
fluorescence   λex 338 nm; λem 375 nm in DMSO

Description

Analysis Note

In addition to the emission maximum at 375 nm, there are lower maxima at 395, 385, 380, 415 and 445 nm.

General description

May contain some tetracene.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
UR2450000
Flash Point(F) 
>392 °F
Flash Point(C) 
>200 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?