• USA Home
  • 82656 - N-(1-Pyrenyl)maleimide

EMAIL THIS PAGE TO A FRIEND
82656 Sigma-Aldrich

N-(1-Pyrenyl)maleimide

for fluorescence, ≥99.0% (HPLC)

Synonym: 1-(1-Pyrenyl)-1H-pyrrole-2,5-dione

  • CAS Number 42189-56-0

  • Empirical Formula (Hill Notation) C20H11NO2

  • Molecular Weight 297.31

  •  Beilstein Registry Number 1542661

  •  EC Number 255-702-1

  •  MDL number MFCD00049301

  •  PubChem Substance ID 329769258

Purchase

Description

Other Notes

Forms fluorescent adducts with proteins and other organic compounds with accessible thiol groups

Application

Reactant used in synthesis of:
• Oligodeoxynucleotide derivatives
• Polymer conjugates of immunogenic peptides
• Modified oligonucleotides for biosensing applications,

Reactant:
• Involved in synthesis of thermally stable fluorescent maleimide/isobutene alternating copolymers
• Used as derivitizing agent for determination of glutathione disulfide levels in biological samples

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P7908 N-(1-Pyrenyl)maleimide

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?