• USA Home
  • 82789 - Pyridine hydrobromide

EMAIL THIS PAGE TO A FRIEND
82789 Sigma-Aldrich

Pyridine hydrobromide

purum, ≥98.0% (AT)

Synonym: Pyridinium bromide

  • CAS Number 18820-82-1

  • Empirical Formula (Hill Notation) C5H5N · HBr

  • Molecular Weight 160.01

  •  Beilstein Registry Number 3615336

  •  EC Number 242-600-7

  •  MDL number MFCD00012801

Purchase

Properties

Related Categories Building Blocks, C5, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   BBFCIBZLAVOLCF-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
impurities   ≤0.5% water
mp   200 °C (dec.)(lit.)
  210-218 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?