• USA Home
  • 82885 - Pyridoxamine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
82885 Sigma-Aldrich

Pyridoxamine dihydrochloride

≥99.0% (TLC)

Synonym: 4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride

  • CAS Number 524-36-7

  • Empirical Formula (Hill Notation) C8H12N2O2 · 2HCl

  • Molecular Weight 241.11

  •  Beilstein Registry Number 3632748

  •  EC Number 208-357-6

  •  MDL number MFCD00012808

  •  PubChem Substance ID 329769267

Purchase

Properties

Related Categories Biochemicals and Reagents, Vitamin B, Vitamin B6, Vitamins
InChI Key   HNWCOANXZNKMLR-UHFFFAOYSA-N
assay   ≥99.0% (TLC)
impurities   ≤8% water
mp   224-226 °C (dec.)(lit.)
storage temp.   −20°C

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
UV1230000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

P9380 Pyridoxamine dihydrochloride, analytical standard, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?