• USA Home
  • 82901 - trans-3-(3-Pyridyl)acrylic acid

EMAIL THIS PAGE TO A FRIEND
82901 Sigma-Aldrich

trans-3-(3-Pyridyl)acrylic acid

purum, ≥98.0% (T)

Synonym: 3-Pyridineacrylic acid

  • CAS Number 19337-97-4

  • Empirical Formula (Hill Notation) C8H7NO2

  • Molecular Weight 149.15

  •  Beilstein Registry Number 113290

  •  MDL number MFCD00006410

  •  PubChem Substance ID 329769268

Purchase

Properties

Related Categories Building Blocks, C8, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   VUVORVXMOLQFMO-ONEGZZNKSA-N
grade   purum
assay   ≥98.0% (T)
mp   232-235 °C (dec.)(lit.)
storage temp.   2-8°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?