• USA Home
  • 82902 - (R)-(+)-α-Methyl-4-pyridinemethanol

EMAIL THIS PAGE TO A FRIEND
82902 Sigma-Aldrich

(R)-(+)-α-Methyl-4-pyridinemethanol

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym: (R)-(+)-1-(4-Pyridyl)ethanol

  • CAS Number 27854-88-2

  • Empirical Formula (Hill Notation) C7H9NO

  • Molecular Weight 123.15

  •  Beilstein Registry Number 4306037

  •  MDL number MFCD00077865

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents, Chiral Resolving Reagents,
InChI Key   HVOAMIOKNARIMR-ZCFIWIBFSA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D +56±3°, c = 1% in chloroform
mp   67-69 °C(lit.)
  67-69 °C

Description

Other Notes

Chiral auxiliary used e.g. in the kinetic resolution of α-arylalkanoic acids

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?