• USA Home
  • 82903 - (S)-(−)-α-Methyl-4-pyridinemethanol

EMAIL THIS PAGE TO A FRIEND
82903 Sigma-Aldrich

(S)-(−)-α-Methyl-4-pyridinemethanol

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym: (S)-(−)-1-(4-Pyridyl)ethanol

  • CAS Number 54656-96-1

  • Empirical Formula (Hill Notation) C7H9NO

  • Molecular Weight 123.15

  •  Beilstein Registry Number 1524527

  •  MDL number MFCD00077866

  •  PubChem Substance ID 24888061

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents, Chiral Resolving Reagents,
InChI Key   HVOAMIOKNARIMR-LURJTMIESA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D −56±3°, c = 1% in chloroform
mp   67-69 °C(lit.)
  67-69 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?