• USA Home
  • 82973 - 4-(2-Pyridylazo)resorcinol monosodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
82973 Sigma-Aldrich

4-(2-Pyridylazo)resorcinol monosodium salt hydrate

purum p.a., for complexometry, ≥98.0% (calc. based on dry substance, UV)

Synonym: PAR-Na

  • CAS Number 16593-81-0

  • Empirical Formula (Hill Notation) C11H8N3NaO2 · xH2O

  • Molecular Weight 237.19 (anhydrous basis)

  •  Beilstein Registry Number 3825637

  •  MDL number MFCD00041025

  •  PubChem Substance ID 329769274

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Indicators, Metal Titration Indicators, Titration More...
grade   for complexometry
InChI Key   KZRPHCQLJZXMJV-IERUDJENSA-M
grade   purum p.a.
assay   ≥98.0% (calc. based on dry substance, UV)
impurities   8-10% water

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

178268 4-(2-Pyridylazo)resorcinol monosodium salt hydrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?