EMAIL THIS PAGE TO A FRIEND
83001 Sigma-Aldrich

(−)-Riboflavin

tested according to Ph.Eur.

Synonym: Lactoflavin, Riboflavinum, Vitamin B2, Vitamin G

  • CAS Number 83-88-5

  • Empirical Formula (Hill Notation) C17H20N4O6

  • Molecular Weight 376.36

  •  Beilstein Registry Number 97825

  •  EC Number 201-507-1

  •  MDL number MFCD00005022

  •  PubChem Substance ID 329769275

Purchase

Properties

Related Categories Analytical/Chromatography, Asymmetric Synthesis, Biochemicals and Reagents, Chemical Synthesis, Chiral Building Blocks,
InChI Key   AUNGANRZJHBGPY-SCRDCRAPSA-N
pharmacopeia   testing conforms to Pharmacopeia
mp   290 °C (dec.)(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?