• USA Home
  • 83215 - Pyrophosphoryl chloride

EMAIL THIS PAGE TO A FRIEND
83215 Sigma-Aldrich

Pyrophosphoryl chloride

purum, ≥97.0% (AT)

Synonym: Diphosphoric tetrachloride, Diphosphoryl chloride, Pyrophosphoric tetrachloride

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Phosphorus Compounds, Phosphorus Halides More...
InChI Key   CNTIXUGILVWVHR-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
refractive index   n20/D 1.476
density   1.82 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3264 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

381829 Diphosphoryl chloride, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?