EMAIL THIS PAGE TO A FRIEND
83310 Sigma-Aldrich

3-Pyrroline

technical, ≥65% (GC)

Synonym: 2,5-Dihydropyrrole

  • CAS Number 109-96-6

  • Empirical Formula (Hill Notation) C4H7N

  • Molecular Weight 69.11

  •  Beilstein Registry Number 103173

  •  EC Number 203-723-1

  •  MDL number MFCD00005213

  •  PubChem Substance ID 24888113

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrrolines
InChI Key   JVQIKJMSUIMUDI-UHFFFAOYSA-N
grade   technical
assay   ≥65% (GC)
  ~30% pyrrolidine basis
refractive index   n20/D 1.46(lit.)
  n20/D 1.46
bp   90-91 °C/748 mmHg(lit.)
density   0.91 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2733PSN1 8(3) / PGII
WGK Germany 
2
Flash Point(F) 
-.4 °F
Flash Point(C) 
-18 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

P75903 3-Pyrroline, 65%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?