EMAIL THIS PAGE TO A FRIEND
83354 Sigma-Aldrich

Mercury(I) chloride

puriss. p.a., ACS reagent, ≥99.5% (RT)

Synonym: Calomel, Mercurous chloride

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Essential Chemicals, M-N, Puriss p.a. ACS,
grade   ACS reagent
InChI Key   ZOMNIUBKTOKEHS-UHFFFAOYSA-L
grade   puriss. p.a.
assay   ≥99.5% (RT)
ign. residue (after reduction)   ≤0.02%
mp   400 °C (subl.)(lit.)
anion traces   sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤10 mg/kg
  HgCl2: ≤100 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 2025 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?