EMAIL THIS PAGE TO A FRIEND
83355 Sigma-Aldrich

Mercury(II) chloride

puriss. p.a., ACS reagent, ≥99.5% (KT)

Synonym: Mercuric chloride

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Essential Chemicals, M-N, Puriss p.a. ACS,
grade   ACS reagent
vapor pressure   1.3 mmHg ( 236 °C)
InChI Key   LWJROJCJINYWOX-UHFFFAOYSA-L
grade   puriss. p.a.
assay   ≥99.5% (KT)
ign. residue (after reduction)   ≤0.02%
mp   277 °C(lit.)
solubility   acetic acid: soluble(lit.)
  acetone: soluble(lit.)
  alcohol: soluble(lit.)
  ethyl acetate: soluble(lit.)
  glycerol: soluble(lit.)
  water: soluble(lit.)
anion traces   sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Price and Availability

Safety & Documentation

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?