EMAIL THIS PAGE TO A FRIEND
83358 Sigma-Aldrich

Mercury(II) cyanide

purum p.a., ≥98.0% (precipitation titration)

Synonym: Mercuric cyanide

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Mercury Salts, Metal and Ceramic Science, Research Essentials,
InChI Key   FQGYCXFLEQVDJQ-UHFFFAOYSA-N
grade   purum p.a.
assay   ≥98.0% (precipitation titration)
density   3.996 g/mL at 25 °C(lit.)
cation traces   Cd: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN 1636 6.1 / PGII
WGK Germany 
3
RTECS 
OW1515000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

208140 Mercury(II) cyanide, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?