EMAIL THIS PAGE TO A FRIEND
83375 Sigma-Aldrich

Mercury(I) iodide

purum p.a., yellow, ≥99.0% (Hg)

Synonym: Mercurous iodide

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Mercury Salts, Metal and Ceramic Science, Research Essentials,
InChI Key   NDKKKEPYKOOXLG-UHFFFAOYSA-L
grade   purum p.a.
assay   ≥99.0% (Hg)
density   7.7 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤500 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Na: ≤100 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability

Safety & Documentation

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?