EMAIL THIS PAGE TO A FRIEND
83377 Sigma-Aldrich

Mercury Orange

≥97.0% (Hg, AAS)

Synonym: 1-(4-Chloromercuriophenylazo)-2-naphthol

Purchase

Properties

InChI Key   CDEIGFNQWMSEKG-QIKYXUGXSA-M
assay   ≥97.0% (Hg, AAS)
storage temp.   2-8°C

Description

Other Notes

For the histochemical and quantitative microdetermination of sulfhydryl groups

Price and Availability

Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?